Learn More
Aviva Systems Biology Corp CDKL2 Antibody - N-terminal region (ARP58606_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58606_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDKL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CDKL2. Alternate names: KKIAMRE, P56ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62373