Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CDKL5, GST Tag Recombinant Protein

Catalog No. A30493
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30493 10 μg
A32864 100 μg
A33353 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A30493 Supplier Invitrogen™ Supplier No. A30493
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This CDKL5 recombinant human protein (1-498) was expressed in insect cells. CDKL5 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic.

CDKL5 serine/threonine kinase (STK9) is associated with X-linked infantile spasm syndrome (ISSX), also known as X-linked West syndrome and Rett syndrome (RTT). CDKL5 can control nuclear speckle morphology by regulating the phosphorylation state of splicing regulatory proteins, and may be involved indirectly in pre-mRNA processing by controlling splicing factor dynamics. The mutations in CDKL5 are major cause of infantile spasms and early epileptic seizures in female patients, and of a later intractable seizure disorder.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O76039
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.25mM DTT, 25% glycerol, 0.1mM PMSF, 0.1mM EDTA, 150mM NaCl, 10mM glutathione and no preservative; pH 7.5
Gene ID (Entrez) 6792
Molecular Weight (g/mol) 82.7 kDa
Name Human CDKL5, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias BC038161; Cdkl5; CFAP247; cyclin dependent kinase 5 transcript; cyclin dependent kinase like 5; cyclin-dependent kinase-like 5; EIEE2; ISSX; RP1-245G19.3; serine/threonine kinase 9; serine/threonine-protein kinase 9; STK9
Common Name CDKL5
Gene Symbol CDKL5
Kinase Family CDKs and CHKs
Protein Length 1-498
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 1-498
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Active, Catalytic
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.