Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci CDKN1C Peptide
SDP

Catalog No. 16594673
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-594-673 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-594-673 Supplier ProSci Supplier No. 7015P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

CDKN1C peptide is used for blocking the activity of CDKN1C antibody.

Specifications

Gene ID (Entrez) 1028
Species Human
Accession Number NP_000067
Common Name cyclin-dependent kinase inhibitor 1 C (p57, Kip2)
Concentration 200 μg/mL
Content And Storage Store CDKN1C peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol CDKN1C
Quantity 0.05 mg
Sequence 17 amino acid peptide near the amino terminus of human CDKN1C.
Synonym BWS, WBS, p57, BWCR, KIP2, p57Kip2, Cyclin-dependent kinase inhibitor 1 C, Cyclin-dependent kinase inhibitor p57
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.