Learn More
Sino Biological CDKN2D / p19ink4d Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 12558MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CDKN2D / p19ink4d (rh CDKN2D / p19ink4d; Catalog#12558-H09E; P55273; Met 10-Leu 166). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7680