Learn More
Aviva Systems Biology Corp CDKN3 Antibody - C-terminal region (ARP53629_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53629_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDKN3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CDKN3. Alternate names: CDI1, CIP2, FLJ25787, KAP, KAP1, MGC70625ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61074