Learn More
Aviva Systems Biology Corp Cdkn3 Antibody - N-terminal region (ARP53628_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53628_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cdkn3. It was validated on Western Blot. . Alternate names: 2410006H10RikApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-860