Learn More
Sino Biological Peptide Substrates: CDKtide
Catalog No. 502569633
Encompass_Preferred
The CDKtide peptide sequence (CKKKYSPTSPSYSPTSPSY-SPTSPS) is derived from the C-terminus of the largest subunit of human RNA polymerase II which has 52 approximate tandem repeats of 7 amino acids Y-[ST]-P-[STQ]-[ST]-P-[SRTEVKGN]. This peptide is suitable as a substrate for CDK7, CDK8, CDK9 and CDK11.
Catalog No. 50-256-9633 Supplier Sino Biological Supplier No. C0658
The CDKtide peptide sequence (CKKKYSPTSPSYSPTSPSY-SPTSPS) is derived from the C-terminus of the largest subunit of human RNA polymerase II which has 52 approximate tandem repeats of 7 amino acids Y-[ST]-P-[STQ]-[ST]-P-[SRTEVKGN]. This peptide is suitable as a substrate for CDK7, CDK8, CDK9 and CDK11.