Learn More
Lonza Walkersville CDM Chondrocyte Basal Medium
Supplier:  Lonza Walkersville CC3226
Encompass
CDM is a chondrocyte cell differentiation basal medium sold separately This can be used with 1 kit of CC-4408 (Chondrocyte Cell Differentiation Medium SingleQuots Supplements and Growth Factors) The full CDM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3225 CDM Chondrocyte Differentiation Medium is specially designed for the differentiation of chondrocytes The BulletKit is conveniently packaged with basal medium and supplements included for the differentiation of chondrocytes CGM Chondrocyte Growth Medium is also available and sold separately for expansion of human chondrocytes
Catalog No. 50-004-96453