Learn More
Lonza Walkersville CDM Chondrocyte Diff Med Sing
Catalog No. 5000496381
Encompass
CDM SingleQuot Kit are components sold separately and does not include the basal medium These are part of the CDM Chondrocyte Differentiation Medium BulletKit (Catalog Number CC-3225) The full CDM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3225 CDM Chondrocyte Differentiation Medium is specially designed for the differentiation of chondrocytes The BulletKit is conveniently packaged with basal medium and supplements included for the differentiation of chondrocytes CGM Chondrocyte Growth Medium is also available and sold separately for expansion of human chondrocytes
Catalog No. 50-004-96381 Supplier Lonza Walkersville Supplier No. CC4408
Add to Cart
Add to Cart
CDM SingleQuot Kit are components sold separately and does not include the basal medium These are part of the CDM Chondrocyte Differentiation Medium BulletKit (Catalog Number CC-3225) The full CDM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3225 CDM Chondrocyte Differentiation Medium is specially designed for the differentiation of chondrocytes The BulletKit is conveniently packaged with basal medium and supplements included for the differentiation of chondrocytes CGM Chondrocyte Growth Medium is also available and sold separately for expansion of human chondrocytes