Learn More
Medchemexpress LLC HY-101455 50mg , CDN1163 CAS:892711-75-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10145550MG
Encompass_Preferred
HY-101455 50mg CDN1163 CAS: 892711-75-0 CDN1163 is an allosteric sarco/endoplasmic reticulum Ca2+-ATPase (SERCA) activator that improves Ca2+ homeostasis. CDN1163 attenuates diabetes and metabolic disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9884