Learn More
Selleck Chemical LLC CDN1163-5mg
SDP
Supplier:  Selleck Chemical LLC S68155MG
Encompass_Preferred
CDN1163 is a small molecular allosteric activator of the sarco/endoplasmic reticulum Ca2+-ATPase (SERCA) that improves Ca2+ homeostasis.
Catalog No. 50-238-9880