Learn More
Medchemexpress LLC 4-(1-methylethoxy)-N-(2-methyl-8-quinolinyl)-benzamide | 892711-75-0 | ≥98.0% | 320.4 | C20H20N2O2 | 5 MG
SDP
Catalog No. 501932137
Encompass_Preferred
CDN1163 is an allosteric activator of sarco/endoplasmic reticulum Ca2+-ATPase (SERCA) used as a research tool to enhance endoplasmic reticulum (ER) calcium uptake and restore cellular Ca2+ homeostasis. It has been reported to reduce peroxide-induced cell death in cultured cells, increase ER calcium uptake in liver microsomes from obese mice, and improve metabolic markers such as blood glucose and triglycerides in animal models.
  • Allosteric activator of SERCA that enhances ER calcium uptake.
  • Improves cellular Ca2+ homeostasis in vitro and in vivo.
  • Reduces peroxide-induced cell death in cultured cells.
  • Improves metabolic parameters in obese mouse models, including reduced blood glucose and triglycerides.
  • High purity suitable for research use (≥98.0%).
  • Available in small-mass pack sizes for dosing flexibility.
Catalog No. 50-193-2137 Supplier Medchemexpress LLC Supplier No. HY1014555MG
CDN1163 is an allosteric activator of sarco/endoplasmic reticulum Ca2+-ATPase (SERCA) used as a research tool to enhance endoplasmic reticulum (ER) calcium uptake and restore cellular Ca2+ homeostasis. It has been reported to reduce peroxide-induced cell death in cultured cells, increase ER calcium uptake in liver microsomes from obese mice, and improve metabolic markers such as blood glucose and triglycerides in animal models.
  • Allosteric activator of SERCA that enhances ER calcium uptake.
  • Improves cellular Ca2+ homeostasis in vitro and in vivo.
  • Reduces peroxide-induced cell death in cultured cells.
  • Improves metabolic parameters in obese mouse models, including reduced blood glucose and triglycerides.
  • High purity suitable for research use (≥98.0%).
  • Available in small-mass pack sizes for dosing flexibility.