Learn More
Biochain Institute Inc cDNA - Dog Normal Tissue: Testis, 40 rxn/PK
SDP
Catalog No. 50179847
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human neurological diseased brain tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-179-847 Supplier Biochain Institute Inc Supplier No. C1734260
BioChain's PCR Ready First Strand cDNAs are available from human neurological diseased brain tissues. 1 ul cDNA is sufficient for each PCR reaction.