Learn More
Biochain Institute Inc cDNA - Dog Normal Tissue: Skeletal Muscle, 40 rxn/PK
SDP
Catalog No. 50179842
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from different dog tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-179-842 Supplier Biochain Institute Inc Supplier No. C1734171
BioChain's PCR Ready First Strand cDNAs are available from different dog tissues. 1 ul cDNA is sufficient for each PCR reaction.