Learn More
Biochain Institute Inc cDNA - Human Adult Normal Tissue: Small Intestine: Ileum, 40 rxn/PK
SDP
Catalog No. 50179951
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from different human adult normal tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-179-951 Supplier Biochain Institute Inc Supplier No. C1234227
BioChain's PCR Ready First Strand cDNAs are available from different human adult normal tissues. 1 ul cDNA is sufficient for each PCR reaction.