Learn More
Biochain Institute Inc cDNA - Human Adult Normal Tissue: Uterus: Fundus, 10 rxn/PK
SDP
Catalog No. 50179971
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from different human adult normal tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-179-971 Supplier Biochain Institute Inc Supplier No. C123427810
BioChain's PCR Ready First Strand cDNAs are available from different human adult normal tissues. 1 ul cDNA is sufficient for each PCR reaction.