Learn More
Biochain Institute Inc cDNA - Human Adult Normal Tissue: Kidney, 40 rxn/PK
SDP
Catalog No. 50179923
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from different human adult normal tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-179-923 Supplier Biochain Institute Inc Supplier No. C1234142
BioChain's PCR Ready First Strand cDNAs are available from different human adult normal tissues. 1 ul cDNA is sufficient for each PCR reaction.