Learn More
Biochain Institute Inc cDNA - Human Adult Normal Tissue: Peripheral Blood Leukocyte, 10 rxn/PK
SDP
Supplier:  Biochain Institute Inc C123414810
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from different human adult normal tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-179-924