Learn More
Biochain Institute Inc cDNA - Human Diabetic Diseased Tissue: Liver, 40 rxn/PK
SDP
Catalog No. 50179984
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human diabetic diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-179-984 Supplier Biochain Institute Inc Supplier No. C1236149DIA
BioChain's PCR Ready First Strand cDNAs are available from human diabetic diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.