Learn More
Biochain Institute Inc cDNA - Human Diabetic Diseased Tissue: Spleen, 40 rxn/PK
SDP
Supplier:  Biochain Institute Inc C1236246DIA
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human diabetic diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-179-992