Learn More
Biochain Institute Inc cDNA - Human Diabetic Diseased Tissue: Esophagus, 40 rxn/PK
SDP
Catalog No. 50179980
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human diabetic diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-179-980 Supplier Biochain Institute Inc Supplier No. C1236106DIA
BioChain's PCR Ready First Strand cDNAs are available from human diabetic diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.