Learn More
Biochain Institute Inc cDNA - Arteriosclerosis Artery and Matched Non-arteriosclerosis Artery, 2 x 10 rxn/PK
SDP
Catalog No. 50180054
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human heart diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-054 Supplier Biochain Institute Inc Supplier No. C8236013HD4PP
BioChain's PCR Ready First Strand cDNAs are available from human heart diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.