Learn More
Biochain Institute Inc cDNA - Coronary Artery Atherosclerosis: Heart, 40 rxn/PK
SDP
Catalog No. 50180045
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human heart diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-045 Supplier Biochain Institute Inc Supplier No. C1236122HD5
BioChain's PCR Ready First Strand cDNAs are available from human heart diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.