Learn More
Biochain Institute Inc cDNA - hypertension: Heart Ventricle, left, 40 rxn/PK
SDP
Catalog No. 50180050
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human heart diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-050 Supplier Biochain Institute Inc Supplier No. C1236138HD2
BioChain's PCR Ready First Strand cDNAs are available from human heart diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.