Learn More
Biochain Institute Inc cDNA - hypertension: Interventricular Septum, 40 rxn/PK
SDP
Catalog No. 50180049
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human heart diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-049 Supplier Biochain Institute Inc Supplier No. C1236130HD2
BioChain's PCR Ready First Strand cDNAs are available from human heart diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.