Learn More
Biochain Institute Inc cDNA - Liver Cirrhosis: Liver, 40 rxn/PK
SDP
Catalog No. 50180064
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human liver diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-064 Supplier Biochain Institute Inc Supplier No. C1236149LCS
BioChain's PCR Ready First Strand cDNAs are available from human liver diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.