Learn More
Biochain Institute Inc cDNA - Liver Cirrhosis: Spinal Cord, 40 rxn/PK
SDP
Catalog No. 50180075
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human liver diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-075 Supplier Biochain Institute Inc Supplier No. C1236234LCS
BioChain's PCR Ready First Strand cDNAs are available from human liver diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.