Learn More
Biochain Institute Inc cDNA - Bronchitis: Lung, 40 rxn/PK
SDP
Catalog No. 50180080
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human lung diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-080 Supplier Biochain Institute Inc Supplier No. C1236152LD2
BioChain's PCR Ready First Strand cDNAs are available from human lung diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.