Learn More
Biochain Institute Inc cDNA - Pneumonia: Lung, 40 rxn/PK
SDP
Catalog No. 50180082
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human lung diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-082 Supplier Biochain Institute Inc Supplier No. C1236152LD4
BioChain's PCR Ready First Strand cDNAs are available from human lung diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.