Learn More
Biochain Institute Inc cDNA - Lupus: Liver: Left Lobe, 40 rxn/PK
SDP
Catalog No. 50180091
Encompass_Preferred
PCR Ready First Strand cDNAs from human lupus diseased tissues are available. More organs maybe available upon request. All the cDNAs are for research use only. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-091 Supplier Biochain Institute Inc Supplier No. C1236150LUP
PCR Ready First Strand cDNAs from human lupus diseased tissues are available. More organs maybe available upon request. All the cDNAs are for research use only. 1 ul cDNA is sufficient for each PCR reaction.