Learn More
Biochain Institute Inc cDNA - Lupus: Spleen, 40 rxn/PK
SDP
Catalog No. 50180098
Encompass_Preferred
PCR Ready First Strand cDNAs from human lupus diseased tissues are available. More organs maybe available upon request. All the cDNAs are for research use only. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-098 Supplier Biochain Institute Inc Supplier No. C1236246LUP
PCR Ready First Strand cDNAs from human lupus diseased tissues are available. More organs maybe available upon request. All the cDNAs are for research use only. 1 ul cDNA is sufficient for each PCR reaction.