Learn More
Biochain Institute Inc cDNA - Human Tumor Tissue: Mesenchymoma, 10 rxn/PK
SDP
Catalog No. 50180182
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human tumor tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-182 Supplier Biochain Institute Inc Supplier No. C123516410
BioChain's PCR Ready First Strand cDNAs are available from human tumor tissues. 1 ul cDNA is sufficient for each PCR reaction.