Learn More
Biochain Institute Inc cDNA - Human Tumor Tissue: Colon, 40 rxn/PK
SDP
Supplier:  Biochain Institute Inc C1235090
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human tumor tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-171