Learn More
Biochain Institute Inc cDNA - Human Tumor Tissue: Liver, 40 rxn/PK
SDP
Catalog No. 50180177
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human tumor tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-177 Supplier Biochain Institute Inc Supplier No. C1235149
BioChain's PCR Ready First Strand cDNAs are available from human tumor tissues. 1 ul cDNA is sufficient for each PCR reaction.