Learn More
Biochain Institute Inc cDNA - Human Tumor Cell Line: A431, 40 rxn/PK
SDP
Catalog No. 50180208
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from tumor cell lines. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-208 Supplier Biochain Institute Inc Supplier No. C1255801
BioChain's PCR Ready First Strand cDNAs are available from tumor cell lines. 1 ul cDNA is sufficient for each PCR reaction.