Learn More
Biochain Institute Inc cDNA - Monkey (Rhesus) Normal Tissue: Stomach: Cardia, 40 rxn/PK
SDP
Catalog No. 50180327
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from different monkey normal tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-327 Supplier Biochain Institute Inc Supplier No. C1534250
BioChain's PCR Ready First Strand cDNAs are available from different monkey normal tissues. 1 ul cDNA is sufficient for each PCR reaction.