Learn More
Biochain Institute Inc cDNA - Mouse Normal Tissue: Stomach, 40 rxn/PK
SDP
Supplier:  Biochain Institute Inc C1334248
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from different mouse normal tissues. 1 ??l cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-356