Learn More
Biochain Institute Inc cDNA - Mouse Normal Tissue: Liver, 40 rxn/PK
SDP
Catalog No. 50180349
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from different mouse normal tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-349 Supplier Biochain Institute Inc Supplier No. C1334149
BioChain's PCR Ready First Strand cDNAs are available from different mouse normal tissues. 1 ul cDNA is sufficient for each PCR reaction.