Learn More
Biochain Institute Inc cDNA - Alzheimer's Disease: Brain: Amygdala, 40 rxn/PK
SDP
Catalog No. 50180105
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human neurological diseased brain tissues. 1 ????l cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-105 Supplier Biochain Institute Inc Supplier No. C1236036ALZ
BioChain's PCR Ready First Strand cDNAs are available from human neurological diseased brain tissues. 1 ????l cDNA is sufficient for each PCR reaction.