Learn More
Biochain Institute Inc cDNA - Alzheimer's Disease: Brain: Postcentral Gyrus, 40 rxn/PK
SDP
Catalog No. 50180149
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human neurological diseased brain tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-149 Supplier Biochain Institute Inc Supplier No. C1236072ALZ
BioChain's PCR Ready First Strand cDNAs are available from human neurological diseased brain tissues. 1 ul cDNA is sufficient for each PCR reaction.