Learn More
Biochain Institute Inc cDNA - Alzheimer's Disease: Brain: Cerebellum, 40 rxn/PK
SDP
Catalog No. 50180108
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human neurological diseased brain tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-108 Supplier Biochain Institute Inc Supplier No. C1236039ALZ
BioChain's PCR Ready First Strand cDNAs are available from human neurological diseased brain tissues. 1 ul cDNA is sufficient for each PCR reaction.