Learn More
Biochain Institute Inc cDNA - Multiple Sclerosis Disease: Brain: Hippocampus, 40 rxn/PK
SDP
Catalog No. 50180124
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human neurological diseased brain tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-124 Supplier Biochain Institute Inc Supplier No. C1236052MSC
BioChain's PCR Ready First Strand cDNAs are available from human neurological diseased brain tissues. 1 ul cDNA is sufficient for each PCR reaction.