Learn More
Andwin Scientific CDNA-PCR BARCOD KIT V14 SQKPCB
SDP
Catalog No. NC3727786
Encompass
SQK-PCB114.24 -cDNA-PCR Barcoding Kit V14.A sequencing kit optimised for identification and quantification of full-length transcripts with highest output including barcodes for up to 24 samples.
Catalog No. NC3727786 Supplier Andwin Scientific Supplier No. 77308578
SQK-PCB114.24 -cDNA-PCR Barcoding Kit V14.A sequencing kit optimised for identification and quantification of full-length transcripts with highest output including barcodes for up to 24 samples.