Learn More
Biochain Institute Inc cDNA - Plant Normal Tissue: cDNA - Plant: Potato, 40 rxn/PK
SDP
Catalog No. 50180371
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from plant tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-371 Supplier Biochain Institute Inc Supplier No. C1634350
BioChain's PCR Ready First Strand cDNAs are available from plant tissues. 1 ul cDNA is sufficient for each PCR reaction.