Learn More
Biochain Institute Inc cDNA - Plant Normal Tissue: cDNA - Plant: Orange, 40 rxn/PK
SDP
Catalog No. 50180370
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from plant tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-370 Supplier Biochain Institute Inc Supplier No. C1634340
BioChain's PCR Ready First Strand cDNAs are available from plant tissues. 1 ul cDNA is sufficient for each PCR reaction.