Learn More
Biochain Institute Inc cDNA - Plant Normal Tissue: cDNA - Plant: Corn, 40 rxn/PK
SDP
Catalog No. 50180369
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from plant tissues. 1 ??l cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-369 Supplier Biochain Institute Inc Supplier No. C1634330
BioChain's PCR Ready First Strand cDNAs are available from plant tissues. 1 ??l cDNA is sufficient for each PCR reaction.