Learn More
Biochain Institute Inc cDNA - Rat Normal Tissue: Brain, 40 rxn/PK
SDP
Catalog No. 50180399
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from different rat normal tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-399 Supplier Biochain Institute Inc Supplier No. C1434035
BioChain's PCR Ready First Strand cDNAs are available from different rat normal tissues. 1 ul cDNA is sufficient for each PCR reaction.