Learn More
Cayman Chemical CdnPMycobacTRIM tubRculo 100ug
SDP
Catalog No. 502994039
Encompass_Preferred
Source: Active recombinant M. tuberculosis strain ATCC 25618/H37Rv N-terminal Trx-, His-, and S-tagged CdnP expressed in E. coli • Amino acids: 1-336 (full length) • MW: 52.71 kDa
Catalog No. 50-299-4039 Supplier Cayman Chemical Supplier No. 22809100 UG
Source: Active recombinant M. tuberculosis strain ATCC 25618/H37Rv N-terminal Trx-, His-, and S-tagged CdnP expressed in E. coli • Amino acids: 1-336 (full length) • MW: 52.71 kDa