Learn More
Sino Biological CDO/CDON Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 106220T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CDO/CDON, and purified by antigen affinity chromatography.
Catalog No. 50-224-8529