Learn More
Enzo Life Sciences CDP-Choline (1g). CAS: 33818-15-4
SDP
Catalog No. 502010561
Encompass_Preferred
Completely inhibits PLA2 activation following cryogenic brain injury in the rabbit at 200 mg/kg. Intermediate in phospholipid biosynthesis. Alternative name: Citicholine sodium. Purity: ≥99%. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-0561 Supplier Enzo Life Sciences Supplier No. BMLEI1231000
Completely inhibits PLA2 activation following cryogenic brain injury in the rabbit at 200 mg/kg. Intermediate in phospholipid biosynthesis. Alternative name: Citicholine sodium. Purity: ≥99%. Solubility: Soluble in water. Long Term Storage: -20°C.